Print…
Goggles, Rafale
Fornecedor: Bollé Safety
BOLERAFARSIEA 9.3 EUR
BOLERAFARSI
Goggles, Rafale
This compact goggle is specially designed to protect against impacts.
  • Compact and robust
  • Adjustable strap
  • Ventilated
Weight: 67 g
Certificações: EN 166 3 4 B (frame) ; 2-1.2 1 B 9 (lens)