Chromatography Data System (CDS) for control of the VWR Hitachi Chromaster and ChromasterUltra Rs.
Functions for: Method creation, data acquisition, chromatograph data processing, DAD data processing and report generation.
PC requirements:
OS: Windows 7 32/64 bit or Windows 10 64 bit
CPU: Intel® Core2 Duo 2 GHz or higher
RAM: 2 GB or more required
Hard disk: At least 2 GB of free space on a partition containing the Windows® 7 system files
Display: Capable of supporting 1280×1024 pixels (256 colours or higher)
Fornecimento: Supplied with software CD with serial number, CD with instruction manuals, installation manual, release note Chromaster System Manager Version 1.1 and software validation certificate.
Atenção: Not 21 CFR Part 11 Compliant, therefore, not suitable for GMP/GLP environments. If 21 CFR Part 11 compliant software is required. Please check DataApex ClarityVA or Agilent Openlab on our webshop.