Print…
Automation reservoirs, Matrix
Fornecedor: Thermo Fisher Scientific
Matrix™
613-2097EA 203 EUR
613-2097 613-2096 613-2099 613-2098 613-2101 613-2100
Automation reservoirs, Matrix
Reservatórios para reagentes
These disposable automation reservoirs made of PP offer excellent chemical resistance. The reservoirs feature the same shape and dimensions as standard microplates, giving users the flexibility to handle reagents on virtually any automated liquid handler.
  • Economical and disposable: No need to wash reservoirs - allowing more reservoirs to be used per run
  • Standard microplate dimensions enables reservoirs to be used in plate handlers or stackers
  • Built-in grooves in reservoir bottom reduce reagent waste and allow consistent aspiration of minimal amounts of reagent
  • Hydrophilic surface so liquid spreads out evenly, with minimal reagent needed to cover bottom
  • Available individually wrapped so reservoirs stay sterile until ready for use; eliminates need to autoclave
Removable splash baffles: Prevents reagents from splashing out when being moved on robotic decks. This enables reservoirs to not only be moved faster, but to be filled closer to the top with reagent. Baffles can be removed to increase the reservoir’s capacity.
Hydrophilic surface properties: Each reservoir undergoes a hydrophilic surface treatment that results in a lessened meniscus, meaning liquid spreads out evenly along the reservoir bottom. Not only does this result in less liquid being needed to cover the reservoir bottom, but it also ensures access to the liquid by all pipette tips.