Print…
Burettes, Pellet pattern, with intermediate stopcock, DAkkS calibration certificate, Schellbach, BLAUBRAND®
Fornecedor: Brand
BRNDDAKKS22767EA 434 EUR
BRNDDAKKS22767
Burettes, Pellet pattern, with intermediate stopcock, DAkkS calibration certificate, Schellbach, BLAUBRAND®
Borosilicate glass 3,3, class AS.
  • Calibrated to deliver 'TD, Ex'
  • Schellbach stripe, ring marks at major graduations
  • Automatic zeroing, intermediate stopcock to recirculate residual liquid
  • Total height of approx 1 m including 2000 ml reservoir bottle
DIN EN ISO 385
Fornecimento: Delivery information: Supplied with 2 litre (soda-lime glass) reservoir bottle
Certificações: DIN EN ISO 385
Informações de encomenda: Burettes are also available on request with an individual certificate or a USP individual certificate.
Fornecimento: Supplied with 2 litre (soda-lime glass) reservoir bottle