1,2-Dimethyl-3-propylimidazolium iodide
Fornecedor: Merck
4.94800.0025EA
0
EUR
4.94800.0025
1,2-Dimethyl-3-propylimidazolium iodide
1,2-Dimethyl-3-propylimidazolium iodide
Fórmula:
C₈H₁₅IN₂ Peso Molecular: 266,1226 g/mol |
Número MDL :
MFCD15072160 Número CAS: 218151-78-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...