Print…
Cleanroom wipes, TechniLite®
Fornecedor: Texwipe
TechniLite®
TEXWTX942EA 0 EUR
TEXWTX942
Cleanroom wipes, TechniLite®
These non woven wipes are a blend of polyester and cellulose. This fabric blend combines the highly absorbent properties of a natural fiber with the cleanliness and strength of a synthetic wipe.
  • Highly absorbent
  • Compatible with most solvents
  • No chemical binders
  • Low extractable levels
Designed for general purpose wiping and spill control in controlled environments.