Gostaria de reiniciar sessão?
Shelving systems, solid, Super Erecta®
Fornecedor: Metro International
Super Erecta®
Perfect for applications involving spillage. Provides barrier between floor and bottom shelf contents to deter contamination.
  • Shelf design features a 3,2 mm raised “ship’s edge” on all four sides to contain spills
  • Louvered/embossed shelf allows air circulation
  • Available in several styles and materials for a variety of applications
  • Shelves are constructed with 18-gauge stainless steel (type 304) or galvanized materials
Standard stainless steel solid shelves (Type 304): with epoxy-coated cast corners address the majority of applications for solid, corrosion resistant shelves or work surfaces. Available in flat solid and louvered/embossed styles.
Galvanized shelves: with uncoated cast corners are ideal for applications requiring a solid shelving or work surface and minimum resistance to corrosion. Available in flat solid and louvered/embossed styles.
Premium Autoclave/Cart Washable Stainless Steel solid shelves (Type 304): feature an all-stainless construction to withstand high temperature, corrosive applications. Available in flat solid style.
ENCOMENDA
Expandir tudo / Esconder tudo
Stainless steel
Descrição Material Peso Und. Referência VWR Quantidade
Super Erecta® solid flat shelf, (width×length): 355×760 mm, 1430FS Aço inoxidável 304 5,4 kg 1 METO1430FS
Super Erecta® solid flat shelf, (width×length): 355×1219 mm, 1448FS Aço inoxidável 304 7,7 kg 1 METO1448FS
Super Erecta® solid flat shelf, (width×length): 355×1524 mm, 1460FS Aço inoxidável 304 9,9 kg 1 METO1460FS
Super Erecta® solid flat shelf, (width×length): 457×610 mm, 1824FS Aço inoxidável 304 5,0 kg 1 METO1824FS
- - - 1 METO1830FS
Super Erecta® solid flat shelf, (width×length): 457×914 mm, 1836FS Aço inoxidável 304 7,3 kg 1 139-0318
Super Erecta® solid flat shelf, (width×length): 457×1066 mm, 1842FS Aço inoxidável 304 8,2 kg 1 139-0319
Super Erecta® solid flat shelf, (width×length): 457×1066 mm, 2436FS Aço inoxidável 304 8,6 kg 1 139-0325
Super Erecta® solid flat shelf, (width×length): 457×1219 mm, 1848FS Aço inoxidável 304 9,1 kg 1 139-0320
Super Erecta® solid flat shelf, (width×length): 457×1524 mm, 1860FS Aço inoxidável 304 10,9 kg 1 METO1860FS
Super Erecta® solid flat shelf, (width×length): 530×914 mm, 2136FS Aço inoxidável 304 8,2 kg 1 139-0321
Super Erecta® solid flat shelf, (width×length): 530×1066 mm, 2142FS Aço inoxidável 304 9,5 kg 1 139-0322
Super Erecta® solid flat shelf, (width×length): 530×1219 mm, 2148FS Aço inoxidável 304 10,4 kg 1 139-0323
Super Erecta® solid flat shelf, (width×length): 530×1524 mm, 2160FS Aço inoxidável 304 11,8 kg 1 139-0324
Super Erecta® solid flat shelf, (width×length): 610×1066 mm, 2442FS Aço inoxidável 304 9,5 kg 1 139-0326
Super Erecta® solid flat shelf, (width×length): 610×1219 mm, 2448FS Aço inoxidável 304 10,9 kg 1 139-0327
Super Erecta® solid flat shelf, (width×length): 610×1524 mm, 2460FS Aço inoxidável 304 14,0 kg 1 139-0328
Super Erecta® solid louvered/embossed shelf, (width×length): 457×610 mm, 1824LS Aço inoxidável 304 5,0 kg 1 METO1824LS
Super Erecta® solid louvered/embossed shelf, (width×length): 457×914 mm, 1836LS Aço inoxidável 304 7,3 kg 1 139-0341
Super Erecta® solid louvered/embossed shelf, (width×length): 457×1066 mm, 1842LS Aço inoxidável 304 8,2 kg 1 139-0342
Super Erecta® solid louvered/embossed shelf, (width×length): 457×1219 mm, 1848LS Aço inoxidável 304 9,1 kg 1 139-0343
Super Erecta® solid louvered/embossed shelf, (width×length): 457×1524 mm, 1860LS Aço inoxidável 304 10,9 kg 1 139-0344
Super Erecta® solid louvered/embossed shelf, (width×length): 530×914 mm, 2136LS Aço inoxidável 304 8,2 kg 1 139-0345
Super Erecta® solid louvered/embossed shelf, (width×length): 530×1066 mm, 2142LS Aço inoxidável 304 9,5 kg 1 139-0346
Super Erecta® solid louvered/embossed shelf, (width×length): 530×1219 mm, 2148LS Aço inoxidável 304 10,4 kg 1 139-0347
Super Erecta® solid louvered/embossed shelf, (width×length): 530×1524 mm, 2160LS Aço inoxidável 304 11,8 kg 1 139-0348
Super Erecta® solid louvered/embossed shelf, (width×length): 610×914 mm, 2436LS Aço inoxidável 304 8,6 kg 1 139-0349
Super Erecta® solid louvered/embossed shelf, (width×length): 610×1066 mm, 2442LS Aço inoxidável 304 9,5 kg 1 139-0351
Super Erecta® solid louvered/embossed shelf, (width×length): 610×1219 mm, 2448LS Aço inoxidável 304 10,9 kg 1 139-0352
Super Erecta® solid louvered/embossed shelf, (width×length): 610×1524 mm, 2460LS Aço inoxidável 304 14,0 kg 1 139-0353
Galvanized
Descrição Material Peso Und. Referência VWR Quantidade
Super Erecta® solid flat shelf, (width×length): 355×914 mm, 1436FG Galvanised steel 6,4 kg 1 METO1436FG
Super Erecta® solid flat shelf, (width×length): 355×1066 mm, 1442FG Galvanised steel 6,8 kg 1 METO1442FG
Super Erecta® solid flat shelf, (width×length): 457×610 mm, 1824FG Galvanised steel 5,0 kg 1 METO1824FG
- - - 1 METO1830FG
Super Erecta® solid flat shelf, (width×length): 457×914 mm, 1836FG Galvanised steel 7,3 kg 1 139-0329
Super Erecta® solid flat shelf, (width×length): 457×1066 mm, 1842FG Galvanised steel 8,2 kg 1 139-0330
Super Erecta® solid flat shelf, (width×length): 457×1219 mm, 1848FG Galvanised steel 9,1 kg 1 139-0331
Super Erecta® solid flat shelf, (width×length): 457×1524 mm, 1860FG Galvanised steel 10,9 kg 1 139-0332
Super Erecta® solid flat shelf, (width×length): 530×914 mm, 2136FG Galvanised steel 8,2 kg 1 139-0333
Super Erecta® solid flat shelf, (width×length): 530×1066 mm, 2142FG Galvanised steel 9,5 kg 1 139-0334
Super Erecta® solid flat shelf, (width×length): 530×1219 mm, 2148FG Galvanised steel 10,4 kg 1 139-0335
Super Erecta® solid flat shelf, (width×length): 530×1524 mm, 2160FG Galvanised steel 11,8 kg 1 139-0336
Super Erecta® solid flat shelf, (width×length): 610×914 mm, 2436FG Galvanised steel 8,6 kg 1 139-0337
Super Erecta® solid flat shelf, (width×length): 610×1066 mm, 2442FG Galvanised steel 9,5 kg 1 139-0338
Super Erecta® solid flat shelf, (width×length): 610×1219 mm, 2448FG Galvanised steel 10,9 kg 1 139-0339
Super Erecta® solid flat shelf, (width×length): 610×1524 mm, 2460FG Galvanised steel 14,0 kg 1 139-0340
Autoclavable and cart-washable solid stainless
Descrição Material Peso Und. Referência VWR Quantidade
Super Erecta® autoclavable and cart-washable solid shelf, (width×length): 457×610 mm, 1824NFS Aço inoxidável 304 5,0 kg 1 METO1824NFS
Super Erecta® autoclavable and cart-washable solid shelf, (width×length): 457×914 mm, 1836NFS Aço inoxidável 304 7,3 kg 1 139-0354
Super Erecta® autoclavable and cart-washable solid shelf, (width×length): 457×1066 mm, 1842NFS Aço inoxidável 304 8,2 kg 1 139-0355
Super Erecta® autoclavable and cart-washable solid shelf, (width×length): 457×1219 mm, 1848NFS Aço inoxidável 304 9,1 kg 1 139-0356
Super Erecta® autoclavable and cart-washable solid shelf, (width×length): 457×1524 mm, 1860NFS Aço inoxidável 304 10,9 kg 1 139-0357
Super Erecta® autoclavable and cart-washable solid shelf, (width×length): 530×914 mm, 2136NFS Aço inoxidável 304 8,2 kg 1 139-0358
Super Erecta® autoclavable and cart-washable solid shelf, (width×length): 530×1066 mm, 2142NFS Aço inoxidável 304 9,5 kg 1 139-0359
Super Erecta® autoclavable and cart-washable solid shelf, (width×length): 530×1219 mm, 2148NFS Aço inoxidável 304 10,4 kg 1 139-0360
Super Erecta® autoclavable and cart-washable solid shelf, (width×length): 530×1524 mm, 2160NFS Aço inoxidável 304 11,8 kg 1 139-0361
Super Erecta® autoclavable and cart-washable solid shelf, (width×length): 610×914 mm, 2436NFS Aço inoxidável 304 8,6 kg 1 139-0362
Super Erecta® autoclavable and cart-washable solid shelf, (width×length): 610×1066 mm, 2442NFS Aço inoxidável 304 9,5 kg 1 139-0363
Super Erecta® autoclavable and cart-washable solid shelf, (width×length): 610×1219 mm, 2448NFS Aço inoxidável 304 10,9 kg 1 139-0364
Super Erecta® autoclavable and cart-washable solid shelf, (width×length): 610×1524 mm, 2460NFS Aço inoxidável 304 14,0 kg 1 139-0365
Gerar códigos de barras Barcode Label Format: 
Criar página do catálogo em PDF
ACESSÓRIOS
Expandir tudo / Esconder tudo
Accessories
Descrição Material Para Und. Referência VWR Quantidade
Clear shelf inlays for Super Erecta® shelving system, 914×355 mm Plástico - 4 Pack METO1436CI-4
Clear shelf inlays for Super Erecta® shelving system, 914×530 mm Plástico - 4 Pack METO2136CI-4
Stationary post, SiteSelect™, stainless steel, 370 mm height - Super Erecta Pro™, Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® solid shelving 1 139-0256
Mobile post, SiteSelect™, chrome plated, 370 mm height - Super Erecta Pro™, Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® solid shelving 1 139-0264
Post mounting shelf supports, width 355 mm, single, chrome plated - - 1 METO1WS14C
Mobile post, SiteSelect™, stainless steel, 370 mm height - Super Erecta Pro™, Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® solid shelving 1 139-0271
Post mounting shelf supports, width 530 mm, single, chrome plated - - 1 METO1WS21C
Poste estacionário, SiteSelect™, cromado, 699 mm de altura - Sistema de estantes Super Erecta® da Drop Mat™, sistema de estantes Super Erecta® muito ajustável, sistema de estantes Super Erecta® maciço, sistema de estantes APPEAL™ 1 139-0250
Decorator shelf inlays, black/white, 610×610 mm - - 1 METO2424BWI
Stationary post, SiteSelect™, stainless steel, 875 mm height - Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving 1 139-0257
Poste móvel, SiteSelect™, cromado, 699 mm de altura - Sistema de estantes Super Erecta® da Drop Mat™, sistema de estantes Super Erecta® muito ajustável, sistema de estantes Super Erecta® maciço, sistema de estantes APPEAL™ 1 139-0265
Post mounting shelf supports, width 355 mm, double, chrome plated - - 1 METO2WS14C
Mobile post, SiteSelect™, stainless steel, 699 mm height - Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving 1 139-0272
Post mounting shelf supports, width 530 mm, double, chrome plated - - 1 METO2WS21C
Additional tabs for rod, bag of 12 - - 12 Pack METO9084Z
Poste estacionário, SiteSelect™, cromado, 875 mm de altura - Sistema de estantes Super Erecta® da Drop Mat™, sistema de estantes Super Erecta® muito ajustável, sistema de estantes Super Erecta® maciço, sistema de estantes APPEAL™ 1 139-0251
Mobile post, SiteSelect™, stainless steel, 857 mm height - Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving 1 139-0273
Stationary post, SiteSelect™, stainless steel, 875 mm height - Super Erecta Pro™ and Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving 1 139-0258
Split sleeves, plastic, white - - 4 Pack METO9985W
Aluminum split sleeves with stainless rings - - 4 Pack METO9986S
Stationary post, SiteSelect™, stainless steel, 1385 mm height - Super Erecta Pro™ and Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving 1 139-0259
Aluminum split sleeves with zinc rings perfect for high temperature or conductive applications ; come with zinc retainer rings to secure the shelf in place (one pack required/shelf) - - 4 Pack METO9986Z
Stationary post, SiteSelect™, chrome plated, 1385 mm height - Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Super Erecta Pro™ shelving 1 139-0252
Poste estacionário, SiteSelect™, Metroseal 3, 1590 mm de altura - Sistema de estantes Super Erecta® muito ajustável, sistema de estantes Super Erecta®, sistema de estantes Super Erecta Pro™ 1 METO63PK3
Clear label holders, plastic, fits 914 mm shelfs, 725×32 mm - - 1 METO9990CL3
Mobile post, SiteSelect™, stainless steel, 1385 mm height - Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Super Erecta Pro™ shelving 1 139-0274
Clear label holders,1520 mm, for 1395×32 mm labels - - 1 METO9990CL5
Mobile post, SiteSelect™, chrome plated, 1385 mm height - Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving 1 139-0267
- - - 1 139-0253
- - - 1 139-0260
Slanted label holders, for 75×32 mm labels - - 1 METO9990SL
Post clamps, joins units together for maximum strength, zinc-plated - - 1 METO9994Z
Poste móvel, SiteSelect™, de aço inoxidável, 1590 mm de altura - Sistema de estantes Super Erecta® muito ajustável, sistema de estantes Super Erecta®, sistema de estantes Super Erecta® maciço, sistema de estantes Super Erecta Pro™ 1 139-0275
- - - 1 139-0268
Poste estacionário, SiteSelect™, Metroseal 3, 1895 mm de altura - Sistema de estantes Super Erecta® muito ajustável, sistema de estantes Super Erecta®, sistema de estantes Super Erecta Pro™ 1 METO74PK3
- - - 1 139-0261
Poste móvel, SiteSelect™, Metroseal 3, 2200 mm de altura - Sistema de estantes Super Erecta® muito ajustável, sistema de estantes Super Erecta®, sistema de estantes Super Erecta Pro™ 1 METO74UPK3
- - - 1 139-0269
Enclosure panel, chrome, 313×1518 mm, for nominal post height 1600 mm - - 1 METOEP36C
- - - 1 139-0254
Enclosure panel, chrome, 464×1791 mm, for nominal post height 1800 mm - - 1 METOEP57C
- - - 1 139-0276
Hanger rail fits on posts along the width or length of the unit 457 mm, fits to size shelf width: 914 mm. Optional hooks can hang from rail at any point. - - 1 METOH118C
Stationary post, SiteSelect™, chrome plated, 191 mm height - Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving 1 139-0248
Mobile post, SiteSelect™, chrome plated, 191 mm height - Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving 1 139-0263
Extension display hanger (152 mm): snaps on Super Erecta® shelf hanger rails and shelf frames, accepts standard carded merchandise - - 1 METOHHK6C
Stationary post, SiteSelect™, stainless steel, 2200 mm height - Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving 1 139-0262
- - - 1 139-0270
- - - 1 139-0277
Stationary post, SiteSelect™, chrome plated, 2454 mm height - Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving 1 METO96P
Swaged post, stainless steel with aluminium cap, 1600 mm dimension - Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta Pro™ shelving 1 METO63UPS-SW
- - - 1 139-0255
Label holders, for 75×32 mm labels - - 1 METO9990P
Plastic split sleeves, black - - 4 Pack METO9985
Slanted label holders, for 75×32 mm labels - - 1 METO9990CL
Storage baskets, chrome plated, 127×345×180 mm, 4 kg - - 1 METOH209C
Storage baskets, chrome plated, 190×440×225 mm, 5 kg - - 1 METOH212C
Gerar códigos de barras Barcode Label Format: 
Criar página do catálogo em PDF
Detalhes
Selecione o artigo para o qual procura o Certificado
Indique o número do lote. Este campo é obrigatório.
Inserir detalhes do certificado
Número de Lote:
Choose from recent batch/lot numbers:
Ou
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Descrição Código de Artigo Disponibilidade Unidade O seu preço Preço por Qtd
Condições Gerais de Venda
Preços
Estas Condições Gerais de Venda anulam automaticamente todas as anteriores.
A VWR International – Material de Laboratório, Lda reserva-se o direito de alterar os preços sem aviso prévio.
Os preços dos produtos indicados serão os preços em vigor no momento da expedição, salvo cotação ou contrato válido.
Todos os preços estão sujeitos a ajustamentos dependentes de quantidades, embalagem especial, custo de matérias-primas ou de produção, expedição em condições especiais, etc.
Despesas de envio
Atendemos com a maior satisfação todos os pedidos, no entanto, reservamo-nos o direito de cobrar despesas de encomenda no valor de 9€ (no Continente) ou 28€ (nas Ilhas) e para encomendas inferiores a 195€ (valor sem IVA), valores de encomenda mínima, de 25€ e 50€ respetivamente.
Para encomendas efetuadas no nosso site, os valores anteriormente mencionados serão reduzidos para 5€ (despesas de encomenda no Continente), 15€ (encomenda mínima) e 47€ para as ilhas, respetivamente.
Relativamente a encomendas de produtos para os quais seja necessário o envio em condições especiais, será cobrado 25€ por despesas de transporte em condições de temperatura controlada e/ou 21,5€ por gastos de transporte especial (Avião). Esta cobrança será feita por cada encomenda.
Aos valores das despesas acima mencionados, acresce o IVA à taxa legal em vigor.
Caso o comprador reclame qualquer isenção, deverá apresentar um comprovativo de isenção válido.
Serviço
O Serviço de atendimento a clientes da VWR processará o seu pedido de uma forma mais rápida sempre que estiver munido das seguintes informações:
  • Nome da Empresa;
  • Número de cliente ou nº de contribuinte;
  • Endereço exato de entrega e de faturação (caso sejam diferentes), incluindo departamento, serviço, email, telefone, etc....);
  • Condições de descarga de mercadoria pesada
  • Códigos dos artigos encomendados de acordo com o catálogo;
  • Número de cotação (caso se aplique);
Para lhe proporcionarmos um melhor serviço e evitar erros, na confirmação por escrito de um pedido previamente efetuado por outra via, terá que, obrigatoriamente mencionar: "pedido previamente por telefone / fax / mail ou internet". A VWR não se responsabiliza por eventuais duplicações de encomendas nos casos em que o cliente não menciona a informação indicada. Qualquer encargo com a devolução do material aos nossos armazéns deverá ser suportado pelo cliente.
Logística e Distribuição
As encomendas serão entregues nos locais indicados na encomenda (de acordo com as condições particulares acordadas) e qualquer transporte a partir deste ponto é da inteira responsabilidade do cliente.
Procuramos entregar sempre os produtos o mais rápido possível. No entanto, todas as datas de entrega são aproximadas e a VWR International – Material de Laboratório, Lda. não poderá ser responsabilizada por qualquer perda ou dano decorrente de qualquer atraso na entrega por razões fora do seu controlo.
Solicitamos que verifique a quantidade e estado exterior das embalagens no ato de receção. Caso verifique qualquer anomalia nas mesmas (caixas danificadas, molhadas ou quantidade de caixas recebidas diferente da mencionada na guia de remessa) esta deverá ser assinalada no comprovativo de receção e reportada ao nosso Serviço de atendimento. Caso não seja feito qualquer registo da anomalia verificada no comprovativo de receção da mercadoria, a VWR não poderá assumir qualquer responsabilidade pelos danos de transporte, ficando todos os encargos a cargo do cliente.
Procedimento de Devoluções
A VWR International – Material de Laboratório, Lda., reserva-se o direito de não aceitar devoluções a não ser em casos excecionais. Todos os pedidos de devolução devem ser solicitados ao nosso Serviço de Atendimento de Clientes, no prazo de até 10 dias da data da guia de remessa. 
Só poderão ser aceites devoluções se os produtos se encontrarem nas suas embalagens originais e em perfeitas condições de revenda. A VWR não se responsabiliza por qualquer devolução que não seja feita mediante a apresentação da nossa guia de recolha por parte da transportadora.
As devoluções de material poderão estar sujeitas a um encargo de rearmazenamento no valor de 10% do valor de faturação (com um valor mínimo de 25€).
A VWR International – Material de Laboratório, Lda. não aceita devoluções de produtos especiais como por exemplo, de transporte e armazenamento em condições de temperatura controlada, produtos químicos ou biológicos de elevada perigosidade, etc. Não serão aceites devoluções efetuadas pelo cliente para os nossos escritórios.
Utilização dos produtos
Os produtos que a VWR fornece são destinados para uso de acordo com a documentação e especificações do fabricante e sob a responsabilidade do usuário. Contudo, para alguns produtos, tais como as susbtâncias ativas, a VWR, como distribuidor, não consegue assegurar o total cumprimento dos requistos legais (European guidelines for GMP medicinal products for human and veterinary use) e não pode vender os produtos em questão para essa finalidade.
Os produtos fornecidos pela VWR não podem ser usados como substâncias ativas para o fabrico de medicamentos de uso humano e veterinário. Para o uso dos produtos da VWR como produto farmacêutico, cosmético, aditivo alimentar e/ou agrícola, pesticida ou para uso doméstico, o cliente deve contatar o serviço local de atendimento da VWR.
O cliente que tenha intenção de utilizar os produtos da VWR como produto farmacêutico, cosmético, aditivo alimentar ou para outro uso para além do recomendado pela VWR devem realizar os seus próprios controlos internos para assegurar que os produtos se encontram em conformidade com a legislação local e diretrizes regulamentares (exemplo: Farmacopeia Europeia, etc...).
Em caso algum, a VWR será responsável se o cliente utilizar um produto numa aplicação diferente da recomendada.
É da responsabilidade do cliente assegurar a conformidade com a regulamentação aplicável (exemplo: a nível de Saúde, Segurança...) e tomar as precauções adequadas relativas ao armazenamento, manuseamento, venda e uso dos produtos.
Condições de Pagamento
Salvo condições específicas previamente acordadas, os preços dados são preços sem IVA com pagamento imediato no ato da encomenda. No caso de o pagamento ser efetuado de forma antecipada, será enviado para o correio electrónico indicado, uma confirmação de encomenda valorizada, sendo que a mesma só será expedida aquando do recebimento do pagamento.
A atribuição de crédito está condicionada à análise de alguns indicadores financeiros, podendo estes, vir a ser solicitados ao cliente.
Para avaliação de outras condições de crédito deverá ser efetuado um pedido por escrito para o email cdc.pt@vwr.com.
O não cumprimento dos prazos de pagamento previamente acordados, poderá ser sujeito a juros de mora, à taxa permitida por lei.
Formas de Pagamento
São aceites pela VWR International - Material de de Laboratório, Lda., os seguintes meios de pagamento: Débito directo, transferência bancária, cheque bancário, vale postal e numerário.
Considera-se como data de pagamento, o dia em que o valor é creditado na conta bancária da VWR International.
Jurisdição Competente
A Jurisdição para todos os litígios resultantes da distribuição de mercadoria e pagamentos é o Tribunal da Comarca de Oeiras.
Matriculada na Conservatóriado Registo Comercial de Cascais
Capital Social: : € 1.084.000
Contrib. Nº 503 842 770
Tel. Geral: + 351-21 3600 770
Fax: +0351-21 3600 798/9
Email: info.pt@vwr.com
https://www.vwr.com/
Print These Terms