Print…
Câmaras digitais, Moticam
Fornecedor: Motic
Moticam®
630-1518EA 0 EUR
630-1518
Câmaras digitais, Moticam
Câmaras
Attachable digital C-mount camera for use in education and laboratories.
  • C-mount connection adapts to most microscopes
  • Microscope specific C-mount to be purchased separately
  • Camera kit includes Ø 30 and 38 mm adapters to fit into the eyepiece tube if no photo port is available
The Moticam 2 camera provides high resolution live imaging up for use in colleges and universities, as well as in clinics and veterinary centres.
Fornecimento: Fornecida com adaptadores de ocular, lâmina de calibração, cabo USB, lente macro (que pode ser focada) e macrotubo para macro-amostras.