Print…
Puncture sets, Plexufix®
Fornecedor: B. Braun
720-2540EA 360 EUR
BRAU4891520
Puncture sets, Plexufix®
Puncture set for 'single shot' technique without nerve stimulation.
  • Consists of plexus needle and connection tubing
  • Needle with 45° short bevel to avoid nerve injury
  • Transparent connection tubing with luer lock hub for immobile needle technique