Print…
Beaker tongs, stainless steel
Fornecedor: Cole Parmer
stuart®
231-0018EA 262 EUR
231-0018 STUAS4/1
Beaker tongs, stainless steel
Pinças /Tongs
These stainless steel tongs with wide jaws (25 mm) firmly grip beakers from 100 to 1500 ml.
  • Jaws are lined with replaceable glass fibre which reduces the risk of breakage
  • With BioCote™ antimicrobial protection
Informações de encomenda: Replacement fibre glass sleeves can be ordered separately.