Print…
Burette clamps, with shaft
Fornecedor: Bochem
BOCH5623EA 27.2 EUR
BOCH5623
Burette clamps, with shaft
Pinças /Clamps Pinças de uso geral
This chromium plated clamp with shaft (length 130 mm) is used for holding burettes (Ø 4 to 20 mm).