Print…
Recirculating coolers, SRC5, SRC14
Fornecedor: Cole Parmer
stuart®
STUASRC5EA 5140 EUR
STUASRC5
Recirculating coolers, SRC5, SRC14
Termóstatos de circulação Sistemas de circulação fechada Circuladores refrigerados
The recirculating coolers offer powerful cooling for an external device. By accurately controlling the temperature of your cooling medium down to -20°C, efficiency of operations such as condensing can be greatly improved. A recirculating cooler is not only much more powerful than conventional water cooling, but is also an ideal alternative when water consumption is an issue for economical, environmental or practical reasons. The SRC5 provides a powerful cooler with a compact footprint suitable for mounting on or under a bench. The LED digital display clearly shows the current temperature of the cooling medium to ±2°C, while the set temperature is revealed by a one button press. The unit has a dedicated drain for easy emptying and cleaning. It also incorporates a built in safety alarm to indicate an overload relay for the refrigeration unit. A dust filter is incorporated and can be accessed without tools via the removable front panel. With all the features of the SRC5 the SRC14 is ideal where larger volumes are required, or when extra cooling capacity is needed, electron microscopes for example. The unit is floor standing with lockable easy roll castors to ensure excellent mobility.
  • Digitally set temperature
  • Easy to read LED display
  • Quiet operation
  • Powerful 450W cooling capacity
  • Safety cut out feature
Max. flow rate / max. pressure:
SRC5: 9 l/min, SRC14: 12 l/min