Print…
5-Mouse anaesthesia kit for small animal imaging systems
Fornecedor: Analytik Jena
UVPA98-0098-04
5-Mouse anaesthesia kit for small animal imaging systems
This anaesthesia system is compatible with the iBox® system.
Non-rebreathing technology safely prevents backflow of gases into the darkroom, virtually eliminating the risk of personnel exposure to anaesthetic gas.
Fornecimento: The 5-mouse anaesthesia unit includes: Isoflurane vaporiser, stand and dual splitter, induction chamber and lid, 3-port mouse rebreather, three mouse nose cones, tabletop charcoal filter stand, charcoal filter canisters, hose for induction chamber and bifurcated fibre optic cables mounted to platform tray. The unit does not include isofluorane, an oxygen cylinder or a pressure regulator. The pressure regulator must have an output pressure of approximately 40-50 psi and fit your gas cylinder.