Print…
Water distillation units
Fornecedor: SI Analytics
171-0034EA 4430 EUR
SCOI285502036 SCOI285502044
Water distillation units
Water distilling apparatus made of DURAN® borosilicate glass 3,3 (hydrolytic class 1 glass) can obtain a distillate of maximum purity that is substantially free from dissolved salts and of extremely low germ content.
  • Easy to clean
  • Cost-effective
The immersion heater is made of high-quality stainless steel so that the presence of heavy-metal ions in the distillate is excluded.
The automatic water level control and built-in boil-dry protection ensure maximum operating safety.
Fornecimento: Supplied with connecting cable, plug and hoses.