Print…
Microbiological safety cabinets, Class II, Safe 2020
Fornecedor: Thermo Scientific
135-1323EA 11332 EUR
HERA51026638
Microbiological safety cabinets, Class II, Safe 2020
Safe 2020 Class II cabinets are designed to deliver maximum safety, comfort and convenience. This biological safety cabinet can be used wherever protection is of paramount importance in safety levels I, II and III, including research, pharmaceutical quality control labs, pharmacies and clinics/hospitals. Combined with the Night-Set-Back mode (reduced blower speed when the window is closed), the Safe 2020 consumes hundreds of kilowatts less energy with lower heat output, than competing cabinets.
  • Superior personnel and product protection with 'best-in-class' airflow and filter technology
  • Exceptionally comfortable ergonomic design prevents user fatigue and promotes safe working habits
  • Maximised energy efficiency for environmental protection and significant cost savings
  • Sash opening: 20 cm working height; can be fully opened to 77,3 cm for thorough cleaning
SmartFlow™ technology maintains constant airflow during normal filter loading or temporary airflow obstruction from foreign objects. The pressure sensor monitoring ensures safe airflow across the entire work surface. An independent alarm system immediately alerts users of any airflow obstruction.
Innovative construction; Its frameless front sash and side glass windows offer an unobstructed view of the complete work area from all angles. Scratch-free, high quality stainless steel work surfaces are easy to clean or autoclave. The indented work modules easily trap spilled liquids. The aerosol-tight window sealing offers additional protection when cabinet is in stand-by mode.
Certificações: These cabinets comply with stringent regulatory standards and are certified to EN 12469.