Tetrapentyltin
Fornecedor: EHRENSTORFER GMBH
EHERC17415100EA
112
EUR
EHERC17415100
Tetrapentyltin
Tetrapentyltin
Fórmula:
C₂₀H₄₄Sn Peso Molecular: 403,28 g/mol |
Número CAS:
3765-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...