Print…
Microplate incubator, SI19
Fornecedor: Cole Parmer
444-0080EA 3400 EUR
444-0080
Microplate incubator, SI19
Incubadoras Incubadoras de Microplacas
The SI19 incubator is designed with laminar flow air circulation and a water tray to provide efficient and uniform heating whilst keeping the humidity high. These features make this equipment suitable for use with up to 24 microplates (48 if stacked in 2 layers) which may be difficult to incubate. An integral over-temperature protection system automatically tracks the set temperature and controls the heater in the event of a fault. Error codes are displayed if a fault has occurred or if the set temperature has been interrupted. Temperature fluctuation and variation ±0,5 °C.
  • Compact with upward opening acrylic door
  • Ideal for microtitre plates and Petri dishes
  • Stainless steel interior
  • Digital setting and LED display of temperature (0,1 °C resolution)
  • Four captive shelves each accommodating six plates