Print…
Heating mantles
Fornecedor: Stuart
454-0060EA 775 EUR
454-0060 454-0061 454-0062 454-0057 454-0058
Heating mantles
Aquecimentos Mantas de aquecimento
Max. 450 °C, with temperature regulation. Range of economical heating mantles with earthed element in case of flask breakage.
  • Fire resistant casing with metal base
  • Low outer case temperatures