Print…
Bottle-top dispensers, Pressmatic PP
Fornecedor: Cole Parmer
stuart®
613-4188EA 0 EUR
613-4188
Bottle-top dispensers, Pressmatic PP
Dispensadores para Frascos
Variable volume, autoclavable.
  • Simple click stop ring for fast and accurate volume setting
  • Clear viewing window to check priming
  • Fully supported delivery tube with drip cap
  • Can be autoclaved at 121 °C
For routine dispensing in the laboratory, the Pressmatic PP offers economy without compromising on performance. Supplied with a factory calibration certificate for guaranteed accuracy the Pressmatic PP is used for dispensing common liquids, dilute acids, buffers, polar solvents, and biological solutions. Based on a PP valve block with glass piston and barrel, the Pressmatic PP is not suitable for dispensing the following liquids: Concentrated or oxidizing acids, e.g. concentrated sulphuric acid and non-polar solvents, e.g. hydrocarbons, halogenated hydrocarbons.
Informações de encomenda: Supplied with three bottle adapters to fit most common laboratory bottles.