Print…
Cool Bag/Cool Box
Fornecedor: J.D. Honigberg International
216-7006EA 519 EUR
216-7006
Cool Bag/Cool Box
Caixas Refrigeradores de bancada
Nylon fabric, PVC coated. Cool bag with biohazard warning.
  • Insulated with 7 mm thick PE foam
  • An insertable partition allows the interior to be subdivided into two different temperature ranges
  • For cooling, two cooling pouches (216-7010 or 216-7013) can inserted into the lid-mounted net
Lightweight bag with adjustable strap. Ideal for transporting temperature-sensitive, biohazardous samples. The bag is closed with a zip fastener. In addition it contains a foam insert with 24 holes for tubes Ø 16 to 20 mm. 