Print…

N-Benzyl-N-methylaniline, Sigma-Aldrich®

Fornecedor: Merck


M-Clarity™ quality level = MQ200

8.40106.0025EA 59.4 EUR
8.40106.0025
N-Benzyl-N-methylaniline, Sigma-Aldrich®
N-Benzyl-N-methylaniline
Fórmula: C₁₄H₁₅N
Peso Molecular: 197,28 g/mol
Densidade: 1,046 g/cm³ (20 °C)
Temperatura de Armazenamento: Ambiente
Número MDL : MFCD00035788
Número CAS: 614-30-2
EINECS: 210-375-4
UN: 1602
ADR: 6.1,III

Encomendar

Especificações dos Resultados do Teste

Solubility 0.009 g/l (20 °C)
Melting Point 8 °C
Molar Mass 197.28 g/mol
Boiling Point 311 °C (1013 hPa)
Flash Point 152 °C
Density 1.046 g/cm3 (20 °C)
Ignition Point 450 °C DIN 51794
Synonyms Benzylmethylphenyl amine
Hill Formula C14H15N

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR