Print…
Rotational viscometers, Alpha
Fornecedor: Fungilab
FUNGV101001EA 3220 EUR
FUNGV101001 FUNGV100002 FUNGV101003 FUNGV101002 FUNGV100001 FUNGV100003
Rotational viscometers, Alpha
Rotational viscometers for quality control in research and development laboratories.
  • Automatic test and automatic range functions
  • Six touch key user interface to navigate the program menu and decide on what is included in the readout
  • Readout with selected speed (rpm), selected spindle, viscosity reading (cP) and selected spindle (SP)
Fornecimento: Supplied with standard spindles (4 (L) and 6 (R and H), stand and spindle protector.