L(-)-Prolina 98.5-101.5% (dried basis) USP, Multi-Compendial, J.T.Baker®
2087-06EA
1219.81
EUR
2087-QM
2087-06
L(-)-Prolina 98.5-101.5% (dried basis) USP, Multi-Compendial, J.T.Baker®
L(-)-Prolina
- Food GMP manufactured product
Fórmula:
C₅H₉NO₂ Peso Molecular: 115,13 g/mol Ponto de ebulição: 245 °C (1013 hPa) Ponto de Fusão: 220…222 °C Densidade: 1,35 g/cm³ (25 °C) Temperatura de Armazenamento: Ambiente |
Número MDL :
MFCD00064318 Número CAS: 147-85-3 EINECS: 205-702-2 Merck Index: 12,07963 |
Especificações dos Resultados do Teste
GMP Manufactured Product | |
Meets B.P. Chemical Specifications | |
Meets E.P. Chemical Specifications | |
Meets U.S.P Requirements | |
Store in well-closed,light-resistant containers | |
Preserve in well-closed containers. | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
USP - Assay (C₅H₉NO₂) (dried basis) | 98.5 - 101.5 % |
USP - Identification | Passes Test |
USP - Specific Rotation [Å]²⁵ ^D | -86.3 - -84.3 Degree |
USP - Loss on Drying at 105°C | ≤0.4 % |
USP - Residue on Ignition | ≤0.4 % |
USP - Chloride (Cl) | ≤0.05 % |
USP - Sulfate (SO₄) | ≤0.03 % |
USP - Iron (Fe) | ≤0.003 % |
USP - Related Compounds - Individual Impurities | ≤0.5 % |
USP - Related Compounds - Total Impurities | ≤2.0 % |
EP/BP - Assay (C₅H₉NO₂) (dried basis) | 98.5 - 101.0 % |
EP/BP - Identification A | Passes Test |
EP/BP - Identification B | Passes Test |
EP/BP - Appearance of Solution | Passes Test |
EP/BP - Specific Rotation [Å]²º^D | -86.0 - -84.0 Degree |
EP/BP-Ninhydrin-Positive Substances-Each | ≤0.2 % |
EP/BP-Ninhydrin-Positive Substances-Total Impurities | ≤0.5 % |
EP/BP - Chloride (Cl) | ≤200 ppm |
EP/BP - Sulfate (SO₄) | ≤300 ppm |
EP/BP - Ammonium (NH₄) | ≤0.02 % |
EP/BP - Iron (Fe) | ≤10 ppm |
EP/BP - Loss on Drying | ≤0.5 % |
EP/BP - Ash (sulfated) | ≤0.1 % |
Endotoxin Concentration, IU/mg, For Information Only | |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Elemental Impurities (USP (232), EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...