Print…
Mid-range UV transilluminator
Fornecedor: Edvotek
733-2006EA 869 EUR
EDVO558
Mid-range UV transilluminator
Designed to visualise DNA stained with ethidium bromide. Safety features include a UV-blocking cover and a power cut-off switch when the cover is opened.