Print…
Rubberfab® Sanitary Lockout/Tagout Tabs
Fornecedor: Rubberfab
MFLX31607-20EA 219.28 EUR
MFLX31607-20 MFLX31607-21 MFLX31607-22
Rubberfab® Sanitary Lockout/Tagout Tabs
Rótulos
Colored tags make for quick and easy identification of process lines.
  • Ideal for lot and batch traceability of your sanitary systems
Lockout/Tagout tabs are available in five colors for quick and easy identification of process lines, resulting in greater safety and efficiency. All lockout/tagouts are numbered for lot and batch traceability. Use in conjunction with the Smart Clamp® sanitary clamps for your complete clamping needs.