Print…
Ultra-low attachment cell culture dishes, Corning®
Fornecedor: Corning
Corning®
CORN4615EA 1355.26 EUR
CORN4615
Ultra-low attachment cell culture dishes, Corning®
Meios de cultura Meios Preparados
Features a covalently bound hydrogel layer that effectively inhibits cellular attachment.
  • Surface minimizes protein absorption, enzyme activation, and cellular activation
  • Surface is non-cytotoxic, biologically inert, and nondegradable
  • Manufactured from optically-clear virgin polystyrene
  • Have stacking beads to aid in handling
The Corning vessels feature a biologically inert ultra-low attachment surface that minimizes cell attachment and promotes the formation of 3D multicellular spheroids employed in cancer research, stem cell biology, and drug screening.
Minimizes cell attachment, protein absorption, and enzyme activation.
Embalagem: 1 case has 8 packs, each pack has 5 plates - totally 40 plates/case.