Print…
Inserts, sterile, Corning® Transwell®
Fornecedor: Corning
Corning®
CORN3467EA 787.04 EUR
CORN3467
Inserts, sterile, Corning® Transwell®
Microplacas Insertos e Placas
Corning® Transwell® microporous membrane inserts are available with a variety of surface treatments for a diverse array of applications
All Transwell® inserts are designed for use in epithelial and endothelial cell culture, transport, migration/invasion, co-culture, and microbial pathogenesis studies.
Inserts provide independent access to both apical and basolateral plasma membranes of the cell monolayer. Sterile.
Tissue culture-treated inserts are available in polyester or polycarbonate formats, and are treated for optimal cell attachment. Once stained, they allow excellent cell visibility using phase contrast microscopy.
Collagen-coated inserts are constructed from polytetrafluoroethylene. Cultrex® basement membrane extract (BME) coated inserts are manufactured from polycarbonate.
Informações de encomenda: Tissue culture-treated inserts are supplied preloaded into multiwell plates. Collagen-coated inserts are individually blister packed, with cluster plates included in each case. The 24-well collagen-coated inserts include twenty-four 6.5 mm inserts packaged separately with two 24-well plates. Cultrex® BME-coated inserts are supplied in two 24-well plates, 12 inserts per plate.