Print…
Adapter Mix II Expansion
Fornecedor: Oxford Nanopore Technologies
OXNTEXP-AMII001EA 200 EUR
OXNTEXP-AMII001
Adapter Mix II Expansion
Reagentes de ácidos nucleicos Next Generation Sequencing Reagents
Vials of Adapter Mix II.
This kit is recommended for users who require additional Adapter Mix II for use with the PCR tiling of SARS-CoV-2 protocols or any other experiments using Native Barcoding Expansions 1-12, 13-24, or 96.
The Adapter Mix II Expansion contain two vials of Adapter Mix II (AMII), sufficient for 12 reactions.
Note: This product is not compatible with kits using Kit 14 (V14) chemistry.
Informações de encomenda: This product is anticipated to be discontinued in September 2023.
Fornecimento: Flow cells and kits are shipped together in an environmentally-friendly temperature-controlled shipping box. Upon receipt, please place the product in a long-term storage location at -20°C. Oxford Nanopore Technologies deem the useful life of the product to be 3 months from receipt by the customer.
Atenção: Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.