Print…
Procurou por: Extracção de proteínas
107  resultados encontrados
SearchResultCount:"107"
Sort Results
Visualização em Lista Easy View (new)
Classifique os resultados da pesquisa
Código de Artigo: (786-247)
Fornecedor: G-Biosciences
Descrição: FOCUS™ Soluble & Insoluble is a complete kit for the selective preparation of soluble (hydrophilic) and insoluble (hydrophobic) proteins from mammalia...
UOM: 1 * 1 KIT
Código de Artigo: (786-250)
Fornecedor: G-Biosciences
Descrição: Lipid rafts are membrane microdomains that are enriched in caveolin, cholesterol, glycolipids, sphingolipids and glycosyl-phosphatidylinositol. Lipid ...
UOM: 1 * 1 KIT
Fornecedor: Thermo Fisher Scientific
Descrição: Pierce™ TMT™ Isobaric Mass Tagging kits and reagents enable multiplex quantitation of proteins extracted from cells and tissues using tandem mass spec...
Fornecedor: G-Biosciences
Descrição: DetergentOUT™ GBS10 resin, DetergentOUT™
Código de Artigo: (PIER90056)
Fornecedor: Thermo Fisher Scientific
Descrição: Chymotrypsin protease is a purified native endoproteinase validated for maximum activity and selectivity in proteomic applications.
UOM: 1 * 100 µG
Código de Artigo: (786-253)
Fornecedor: G-Biosciences
Descrição: FOCUS™ Glycoprotein kit is based on lectin binding of specific glycoproteins with terminal α-D mannosyl and α-D glycosyl proteins.
UOM: 1 * 1 KIT
Código de Artigo: (786-249)
Fornecedor: G-Biosciences
Descrição: FOCUS™ Membrane Proteins for the fractionation of highly enriched membrane proteins.
UOM: 1 * 1 KIT
Fornecedor: Biotium
Descrição: AccuOrange™ Protein Quantitation Kit is a highly sensitive fluorescence-based assay for quantitating purified protein samples in 96-well format (excit...
Fornecedor: G-Biosciences
Descrição: Antibody Fab Fragmentation Kits are designed for the generation and isolation of Fab fragments from IgG molecules. The kits utilise Immobilised Papain...
Fornecedor: Thermo Fisher Scientific
Descrição: The Pierce™ Universal Nuclease for Cell Lysis is ideal for a wide variety of applications where complete digestion of nucleic acids is needed when pre...
Fornecedor: G-Biosciences
Descrição: The use of peptides for the generation of antibodies against specific peptides has become an essential tool in proteomic research. A crucial step in u...
Código de Artigo: (786-022)
Fornecedor: G-Biosciences
Descrição: FOCUS™ Mitochondria kit is specifically designed for the isolation of intact mitochondria from cultured mammalian cells. This kit allows for the fast ...
UOM: 1 * 1 KIT
Código de Artigo: (PIER90054)
Fornecedor: Thermo Fisher Scientific
Descrição: Glu-C protease is a highly specific endoproteinase use to improve sequence coverage in mass spectrometry protein identification applications.
UOM: 1 * 50 µG
MSDS
Código de Artigo: (786-598)
Fornecedor: G-Biosciences
Descrição: Immobilised biotin resin is designed for the high affinity chromatography purifications of avidin, streptavidin and neutravidin protein. The resin con...
UOM: 1 * 5 mL
Código de Artigo: (786-241)
Fornecedor: G-Biosciences
Descrição: KIT INGEL SILVER 100 PREPS 1 * 1 KIT
UOM: 1 * 1 KIT
Fornecedor: Thermo Fisher Scientific
Descrição: B-PER™ Complete Bacterial Protein Extraction Reagent is an all-in-one formulation of non ionic detergent and enzymes that enables mild extraction of p...
Preço sob consulta
O stock para este item é limitado. Por favor, certifique-se de que efetuou o seu login para visualizar o stock disponível. Se a call ainda é exibida e precisar de ajuda, por favor, ligue para 213 600 770
O stock para este item é limitado. Por favor, certifique-se de que efetuou o seu login para visualizar o stock disponível. Se a call está visível e precisar de ajuda, por favor, ligue para 213 600 770
Este produto é sujeito a regulamentação especifica.
Em caso de encomenda, será contactado a solicitar documentação complementar necessária e/ou obrigatória (licença, autorização ou declaração de uso final) para a continuidade do pedido. Agradecemos a vossa compreensão
Este produto é sujeito a regulamentação especifica.
Em caso de encomenda, será contactado a solicitar documentação complementar necessária e/ou obrigatória (licença, autorização ou declaração de uso final) para a continuidade do pedido. Agradecemos a vossa compreensão.
Este produto está bloqueado. Para obter mais informações, contacte a VWR através do número 213 600 770.
O produto pretendido já não se encontra disponível. O produto indicado é um substituto.
Este produto encontra-se em rutura de stock. Poderá encontrar alternativas pesquisando pelo código de artigo indicado acima. Se precisar de ajuda, por favor contacte a VWR através do 213 600 770.
1 - 16 of 107
no targeter for Bottom