Print…
Automated plate washing for ELISA, cells and beads
Improve lab efficiency & optimize results with fast and efficient plate washers. Compare and find the optimal plate washer for your applications.
Compare now
TECAN microplate washers
Learn more
Procurou por: Ensaios
651  resultados encontrados
SearchResultCount:"651"
Sort Results
Visualização em Lista Easy View (new)
Classifique os resultados da pesquisa
Fornecedor: SIGMA-ALDRICH MICROSCOPY
Descrição: Bicinchoninic acid is used for the determination of protein concentration. It is responsible for the making of intense purple complex with cuprous ion...
Código de Artigo: (GAHK20-1KT)
Fornecedor: Merck
Descrição: The Glucose (HK) Assay Kit is for the quantitative, enzymatic determination of glucose in food and other material.
UOM: 1 * 1 unid.
Código de Artigo: (M172-1L)
Fornecedor: VWR Chemicals
Descrição: The Bradford protein assay is the preferred colorimetric assay for quantifying total protein concentration. Based upon complex formation between basic...
UOM: 1 * 1 L
MSDS Certificados
Código de Artigo: (1B1301-KIT)
Fornecedor: VWR Chemicals
Descrição: The Protein Precipitation Kit utilises a simple trichloroacetic acid (TCA) based method to precipitate proteins in complex mixtures. Only protein in t...
UOM: 1 * 1 KIT
Código de Artigo: (1B1558-KIT)
Fornecedor: VWR Chemicals
Descrição: Fluorescent DNA Quantitation Kit uses fluorescent bisBenzimide H33258 dye to bind to AT sequences of the minor groove of double-stranded DNA. In the a...
UOM: 1 * 1 KIT
Fornecedor: Biotium
Descrição: CellBrite kits are designed to label cell cytoplasmic membranes with a fluorescent membrane dye pre-dissolved as an optimised staining solution. By co...
Fornecedor: Tonbo Biosciences
Descrição: Ghost Dye solutions allow for clear live vs. dead cell discrimination for more precise flow cytometry analysis, and are especially useful for intracel...
Fornecedor: Biotium
Descrição: MTT, XTT, and resazurin are reduced by mitochondrial metabolic activity to yield coloured or fluorescent products, and thus are useful for and assayin...
Fornecedor: Biotium
Descrição: Firefly luciferase is widely used as a transcriptional reporter gene for research and drug screening. The Steady-Luc™ HTS kit can be used with lumines...
Fornecedor: Biotium
Descrição: ATP-Glo™ Bioluminometric Cell Viability Assay offers a highly sensitive assay for quantifying ATP. The homogeneous assay involves a single addition of...
Fornecedor: Biotium
Descrição: Preservative-free Annexin V conjugates are compatible with detection of apoptotic cells in live cell imaging experiments, fluorescence microscopy, or ...
Fornecedor: Biotium
Descrição: Cellular assay, SynaptoRed™ C2 (FM4-64), m=1, n=3
Fornecedor: Biotium
Descrição: CF® dye labeled dextrans could be used as a fluorescent fluid-phase markers to study cell permeability, endocytosis, or mechanisms of biomolecular del...
Fornecedor: Biotium
Descrição: RedDot™1 is a far-red cell membrane-permeable nuclear dye similar to Draq5™. The dye is ideal for specifically staining the nuclei of live cells.
Fornecedor: CELL VIABILITY INC.
Descrição: The Cell Viability Reagent Kits are designed specifically for use with the Beckman Coulter Vi-CELL® Cell Viability Analyser. The Cell Viability Reagen...
Fornecedor: Biotium
Descrição: AccuGreen™ kit is specific for dsDNA and can quantify DNA samples in the range of 0,1 to 10 ng/μl.
Preço sob consulta
O stock para este item é limitado. Por favor, certifique-se de que efetuou o seu login para visualizar o stock disponível. Se a call ainda é exibida e precisar de ajuda, por favor, ligue para 213 600 770
O stock para este item é limitado. Por favor, certifique-se de que efetuou o seu login para visualizar o stock disponível. Se a call está visível e precisar de ajuda, por favor, ligue para 213 600 770
Este produto é sujeito a regulamentação especifica.
Em caso de encomenda, será contactado a solicitar documentação complementar necessária e/ou obrigatória (licença, autorização ou declaração de uso final) para a continuidade do pedido. Agradecemos a vossa compreensão
Este produto é sujeito a regulamentação especifica.
Em caso de encomenda, será contactado a solicitar documentação complementar necessária e/ou obrigatória (licença, autorização ou declaração de uso final) para a continuidade do pedido. Agradecemos a vossa compreensão.
Este produto está bloqueado. Para obter mais informações, contacte a VWR através do número 213 600 770.
O produto pretendido já não se encontra disponível. O produto indicado é um substituto.
Este produto encontra-se em rutura de stock. Poderá encontrar alternativas pesquisando pelo código de artigo indicado acima. Se precisar de ajuda, por favor contacte a VWR através do 213 600 770.
1 - 16 of 651
no targeter for Bottom