Print…
Bacillus circulans 1,3-alpha-Glucosidase 87A
Fornecedor: Apollo
APOSBIZY1031-250UGEA 281 EUR
APOSBIZY1031-250UG
Bacillus circulans 1,3-alpha-Glucosidase 87A
1,3-alpha-Glucosidases participate in the endohydrolysis of 1,3-alpha-D-glucosidic linkages in isolichenin,pseudonigeran and nigeran. Family: GH87. E.C.:3.2.1.59. Architecture: GH87. Main Activity:glucan endo-1,3-alpha-glucosidase