2,4'-DDT
Fornecedor: Merck
|
Perigo
|
49018.EA
271
EUR
49018.
2,4'-DDT
2,4'-DDT
Fórmula:
C₁₄H₉Cl₅ Peso Molecular: 354,49 g/mol |
Número CAS:
789-02-6 EINECS: 212-332-5 UN: 2761 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...