6-Fluoro-3-formylchromone ≥98.0% (by GC, titration analysis)
Fornecedor: TCI EUROPE
|
Atenção
|
TCIAF0905-1GEA
25
EUR
TCIAF0905-1G
6-Fluoro-3-formylchromone ≥98.0% (by GC, titration analysis)
6-Fluoro-3-formylchromone
Fórmula:
C₁₀H₅FO₃ Peso Molecular: 192,15 g/mol |
Número MDL :
MFCD00139060 Número CAS: 69155-76-6 |
Especificações dos Resultados do Teste
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 155.0 to 159.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...