Print…

Disodium metasilicate nonahydrate 90-100%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Perigo

3868-01EA 163.52 EUR
3868-01 3868-05 EXPOJT3868-1
Disodium metasilicate nonahydrate 90-100%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Disodium metasilicate nonahydrate

Crystals. Lot analysis on label.

Fórmula: Na₂SiO₃·9H₂O
Peso Molecular: 284,2 g/mol
Ponto de Fusão: 40 °C
Densidade: 2,62 g/cm³ (21 °C)
Número CAS: 13517-24-3
EINECS: 229-912-9
UN: 3253
ADR: 8,III

Encomendar

Especificações dos Resultados do Teste

For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Assay, For Information Only
Chloride (Cl) ≤0.01 %
Sulfate (SO₄) ≤0.01 %
Heavy Metals (as Pb) ≤0.001 %
Iron (Fe) ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR