Print…
Sign and label printer, BradyPrinter S3000
Fornecedor: Brady
BRDY241114EA 1680 EUR
BRDY241115 BRDY241114 BRDY149513 BRDY149512
Sign and label printer, BradyPrinter S3000
Impressoras Label Printers
Easily print reliable signs for more safety and efficiency.
  • Continuous tape and die cut labels
  • Monocolour colour capability
With the BradyPrinter S3000, you can print a wide range of signs and labels to increase safety and efficiency in any workplace. Safety signs, floor marking, 5S labels, lean labels, labels to identify racks, doors, stairs, bins, desks and chairs and CLP-labels to identify chemicals can all be printed in monocolour at your location whenever you need them.
The BradyPrinter S3000 prints on Brady’s industrial grade vinyls that offer 8-10 year outdoor durability, and on polyesters including ToughStripe floormarking. Signs and labels printed with the BradyPrinter S3000 can be used both in- and outdoor, which is extremely useful for safety and facility identification in large plants, in and around production halls, warehouses and logistics and distribution centres. Brady’s sign &label materials are designed to last in industrial environments and have been tested in line with international ASTM test methods. Technical data sheets are available for every sign and label material.
The BradyPrinter S3000 is extremely easy to use and features autocalibration and automatic label material setup. This will enable you to just walk up to the printer and start printing the signs and labels you
need. Switching to another label material is easy and extremely fast thanks to the printer’s ‘drop-in’ label materials and matching ink ribbons that can be switched out in less than 20 seconds. When used with Brady Workstation apps, a wide variety of sign and label design options becomes available.
Certificações: UL, cUL, CE, RoHS, RCM, EAC, KCC, BIS, BSMI, Taiwan RoHS, CCC, China RoHS.
Fornecimento: Includes an S3000 Printer, a product CD, a power cord, a Brady Workstation Suite, a USB cable, a cleaning kit, a cutter cleaning tool and a quick start guide.