Print…
Cleaning swabs, sealed foam, CONSTIX® SF-11
Fornecedor: Contec
CONTSF-11_UEA 179 EUR
CONTSF-11_U
Cleaning swabs, sealed foam, CONSTIX® SF-11
The SF-11 swab consists of a foam head formed over the end of a 6” cotton-tipped applicator with wood handle. The foam head is an 100 PPI sealed closed-cell polyurethane foam.
  • Birch hardwood handle makes it stiff enough for vigorous swabbing
  • Wood handle is solvent resistant
  • Soft polyurethane foam head provides good particle entrapment and removal
  • No adhesives are used
The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily absorbed for applications involving solvent delivery or removal. The swab is low in particles and fibers and is well suited to applying solutions and adhesives.
Though compatible with many common solvents, the SF-11 is not recommended for use with ketones such as acetone or MEK.
Embalagem: Swabs are packaged 500 per case.