Print…
Cleaning swabs, sealed, CONSTIX® SP-11
Fornecedor: Contec
CONTSP-6MEA 217 EUR
CONTSP-6M
Cleaning swabs, sealed, CONSTIX® SP-11
The SP-6 sealed swab consists of 100% knitted microfiber polyester fabric that is thermally bonded to a flex-tip PP handle.
  • Durable and absorbent
  • Feature extremely low particles and residue levels
  • Flexible tip provides a softer application on surfaces
  • Polyester swab head provides enhanced sorbency, particle entrapment, and removal
  • Flexible tip of the SP-6M swab provides a softer application of the swab head to surfaces
The swab head is made of cleanroom laundered; double-layer knitted microfibre polyester material. The swab head is constructed without adhesives or binders, making it compatible with most common solvents, such as alcohols and ketones.
Embalagem: Swabs are packed 500 per case.
Atenção: * O (s) produto (s) nesta página não se enquadra no âmbito da Diretiva da UE 2019/904