Print…
Disposable mop covers, Clino® one way tech
Fornecedor: Pfennig
PPSA3500323EA 16.8 EUR
PPSA3500323
Disposable mop covers, Clino® one way tech
These mop covers are a cost-effective, non-sterile alternative for cleanroom purposes. These are very high quality, clean room suitable processing type.
  • Low particle opening system
  • vacuum-packed
  • Washed and packed in the clean room
  • Made of 100% polyester with back lining
For all technical clean rooms and non-sterile GMP areas and most ideal to be used on all common cleanroom and pharmaceutical floors. This provides high and maximum cleaning performance with minimal particle release.
Certificações: Up to ISO purity class 6; Up to GMP cleanliness class C/D.
Embalagem: Five pieces double packed, 20 pieces/carton.