Print…
Ashing furnaces with lift up door, Tmax 1100 °C and standard controller B510, LVT 3/11 - LVT 15/11
Fornecedor: Nabertherm
NABELV034H1LN1EA 0 EUR
NABELV034H1LN1 NABELV094H1LN1 NABELV154H1LN1TZ NABELV094H1LN1TZ NABELV034H1LN1Z NABELV094H1LN1T NABELV054H1LN1TZ NABELV094H1LN1Z NABELV054H1LN1Z NABELV154H1LN1T NABELV054H1LN1T NABELV034H1LN1T NABELV034H1LN1TZ NABELV154H1LN1Z NABELV054H1LN1 NABELV154H1LN1
Ashing furnaces with lift up door, Tmax 1100 °C and standard controller B510, LVT 3/11 - LVT 15/11
The LVT 3/11 to LVT 15/11 ashing furnaces are specially designed for ashing in the laboratory. A special air intake and exhaust system allows air exchange greater than six times per minute. Incoming air is preheated to ensure good temperature uniformity.
  • Heating from two sides
  • Ceramic heating plates with integral heating element, which is protected from fumes and splashing, and easy to replace
  • Exclusive use of insulation materials without categorization according to EC Regulation No 1272/2008 (CLP). This explicitly means that alumino silicate wool, also known as “refractory ceramic fibre” (RCF), which is classified and possibly carcinogenic, is not used
  • Lift up door (LVT) with hot surface facing away from the operator
  • Good temperature uniformity due to preheating incoming air
  • Air exchange greater than six timer per minute
The furnace is equipped with the B510 controller with touch screen display, featuring 5 freely adjustable programs and 4 segments and 2 extra functions. These controllers are PID microprocessor controlled with a self-diagnostic system and integrated process documentation.
Minutes to Tmax without load and standard voltage: 120
Informações de encomenda: Optional accessories such as over-temperature limit controller with adjustable temperature cut out for thermal protection Class 2, in accordance with EN 60519-2 as temperature limiter to protect the oven and load, access port for thermocouple. Corresponding models with fold down doors, different Tmax and other controllers are also available. Please search the website for more details.
Atenção: Maximum continuous operating temperature should be 50 °C below the 'Max. temp.' of the model shown in the table above.
Outer dimensions include exhaust tube (Ø 80 mm).