Print…
Ashing furnaces with fold down door, Tmax 1100 °C and standard controller B510, LV 3/11 - LV 15/11
Fornecedor: Nabertherm
NABELV034K1LN1TZEA 0 EUR
NABELV034K1LN1TZ NABELV152K1RN1TZ NABELV034K1LN1Z NABELV154K1LN1TZ NABELV054K1LN1 NABELV052K1RN1TZ NABELV154K1LN1 NABELV154K1LN1Z NABELV032K1RN1TZ NABELV094K1LN1TZ NABELV092K1RN1TZ NABELV094K1LN1T NABELV034K1LN1 NABELV092K1RN1T NABELV154K1LN1T NABELV094K1LN1Z NABELV052K1RN1T NABELV152K1RN1Z NABELV032K1RN1T NABELV034K1LN1T NABELV152K1RN1T NABELV052K1RN1Z NABELV032K1RN1Z NABELV092K1RN1Z NABELV094K1LN1
Ashing furnaces with fold down door, Tmax 1100 °C and standard controller B510, LV 3/11 - LV 15/11
The LV 3/11 to LV 15/11 ashing furnaces are specially designed for ashing in the laboratory. A special air intake and exhaust system allows air exchange greater than six times per minute. Incoming air is preheated to ensure good temperature uniformity.
  • Heating from two sides
  • Ceramic heating plates with integral heating element which is protected from fumes and splashing, and easy to replace
  • Exclusive use of insulation materials without categorization according to EC Regulation No 1272/2008 (CLP). This explicitly means that alumino silicate wool, also known as “refractory ceramic fibre” (RCF), which is classified and possibly carcinogenic, is not used
  • Fold down door (LV) which can be used as work platform
The furnace is equipped with the B510 standard controller with touch screen display, featuring 5 freely adjustable programs and 4 segments and 2 extra functions. These controllers are PID microprocessor controlled with a self-diagnostic system and integrated process documentation.
Minutes to Tmax without load and standard voltage: 120
Informações de encomenda: Optional accessories such as over-temperature limit controller with adjustable temperature cut out for thermal protection Class 2, in accordance with EN 60519-2 as temperature limiter to protect the oven and load, access port for thermocouple. Corresponding models with lift up doors, different Tmax and other controllers are also available. Please search the website for more details.
Atenção: Maximum continuous operating temperature should be 50 °C below the 'Max. temp.' of the model shown in the table above.
Outer dimensions include exhaust tube (Ø 80 mm).