Print…
Waste bins, self extinguishing
Fornecedor: asecos
MSGA23148EA 56 EUR
MSGA23148 MSGA23155 MSGA23151 MSGA23153 MSGA23143
Waste bins, self extinguishing
These self extinguishing waste bins provide fire protection in work areas where easily flammable waste is disposed of.
  • Available in choice of colours and finishes
  • Smooth and shiny surface
  • Corrosion-protected
Container and extinguishing attachment are made of stainless steel.