These high-temperature furnaces are used to determine combustion loss during annealing and for thermogravimetric analysis (TGA) in the laboratory.
- Defined application within the constraints of the operating instructions
- Has a scale for measuring weight reduction during annealing and with gas supply system
Process control and documentation for temperature and combustion loss via VCD software package for monitoring, documentation and control.
Fornecimento: Complete system includes the high-temperature furnace for use at 1600 or 1750 °C, a table frame, precision scale with feedthroughs into the furnace and powerful software for recording both the temperature curve and the weight loss over time.