Print…
Paraffin wax dispenser, Electrothermal
Fornecedor: Cole Parmer
Electrothermal
114-8737EA 3680 EUR
114-8737
Paraffin wax dispenser, Electrothermal
Paraffin Wax Trimmers and Dispensers
The Electrothermal Paraffin Wax Dispenser delivers on-demand molten wax for easy and mess-free tissue infiltration and embedding. It provides convenient 'wax on tap' via a heated tap which prevents blockages and ensures an even flow of wax on-demand for Histology and Pathology laboratories. The Paraffin Wax Dispenser is designed for melting up to 6 kg of pelletised wax at a time, and has a large 7,5 L capacity tank.
  • Ambient to 70 °C operating temperature range
  • Silver based anti-microbial coating to inhibit growth of bacteria
  • Safety features include over-temperature cut-out at 120 °C
  • Heated tap to prevent blockages
The slimline design of the instrument ensures that it takes up minimal bench space in the laboratory. The Wax Dispenser has digital temperature control which provides accurate temperature control of the molten wax, and is coupled with an ultra-fast heating system for rapid melting of pelletised wax. A non-drip lever tap with three settings (off, on-demand pull dispense and continuous flow) delivers the wax on-demand. The inner tank has a filter screen (0.5 mm mesh) fitted to prevent coarse particles from blocking the delivery tap.
An intuitive digital control panel allows control of the Wax Dispenser. The set temperature can be controlled using the up and down arrows on the control panel. The wax dispenser is designed to melt large quantities of pelletised wax as fast as possible. When set at 65 ˚C the instrument will take ~90 minutes to fully melt 5 kg of pelletised wax, with quantities of molten wax available for use in 30 minutes.
The equipment is protected by a miniature circuit breaker and a temperature cut out. If an over-temperature condition occurs, the Wax Dispenser will automatically turn off at 120 °C.