Print…
ESD safety shoes, lace-up, Ergo-Active, Rob
Fornecedor: Elten
ELTN7247102/41EA 0 EUR
ELTN7247102/41 ELTN7247102/42 ELTN7247102/43 ELTN7247102/44 ELTN7247102/45 ELTN7247102/46 ELTN7247102/47 ELTN7247102/48 ELTN7247102/40
ESD safety shoes, lace-up, Ergo-Active, Rob
ESD safety shoes made from hydrophobised cowhide leather/textile material, leather lining, a full padded bellows tongue plus a dual-density TPU-PU sole (Ergo-Active) with TPU pivots.
  • Leather lining
  • Full length inlay sole (Ergo-Active ESD)
  • Scuff cap protects leather upper
  • Metal-free puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Black/grey.
Certificações: EN 61340 (ESD); EN ISO 20345; S3, SRC.