Print…
Heat transfer liquids, Ultra 240
Fornecedor: Lauda
461-2846EA 1030 EUR
LAUDLZB108 LAUDLZB208 LAUDLZB308
Heat transfer liquids, Ultra 240
These transfer fluids allow highly accurate thermostating, even at extreme temperatures.
  • Durability
  • Simple and safe handling
  • Reliability, suitable for long-term operation
Recommended temperature range: 80 to 240 °C (open system).
* Denotes silicone oil