Print…
Whole blood, breast cancer (BC)
HCMSPB000F-2-BCEA 0 EUR
HCMSPB000F-2-BC
Whole blood, breast cancer (BC)
Whole blood used for researching the causes, diagnosis, supportive care and treatment of breast cancer (BC).The whole blood is collected from IRB-consented donors diagnosed with breast cancer (BC).
No pre screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2.
Storage: Use immediately upon receipt.
Atenção: Intended for only research use and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.