Gostaria de reiniciar sessão?
Human serum, psoriatic arthritis (PA)
Fornecedor: HemaCare
PB007C-1-PAEA 0 EUR InStock
PB007C-1-PA
Human serum, psoriatic arthritis (PA)
Soro
Human serum used for researching the causes, diagnosis, supportive care and treatment psoriatic arthritis (PA). Collected from IRB-consented donors diagnosed with PA.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2, HTLV-I/II, HIV-1/HCV/HBV nucleic acid, WNV nucleic acid, Trypanasoma cruzi and Syphilis.
Storage: Store at −80 °C.
Atenção: Intended for research use only and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.
Order Now
ENCOMENDA
Inserir detalhes do certificado
Número de Lote:
Choose from recent batch/lot numbers:
Ou
Review & Compare Alternatives
Original Product:
Descrição Código de Artigo Disponibilidade Unidade O seu preço Preço por Qtd
Capital Social: : € 1.084.000
Contrib. Nº 503 842 770
Tel. Geral: + 351-21 3600 770
Fax: +0351-21 3600 798/9
Email: info.pt@vwr.com
https://www.vwr.com/
Print These Terms