Print…
RNA system, QuantiFluor®
Fornecedor: Promega
PROME3310EA 603 EUR
PROME3310
RNA system, QuantiFluor®
Ensaios Nucleic Acid Assays
The QuantiFluor® RNA system contains a fluorescent RNA-binding dye that enables sensitive quantitation of small amounts of RNA in solution.
  • Highly sensitive and easy-to-use
  • Requires less template RNA (100 pg) than other systems
  • More sensitive than absorbance methods for low-concentration samples
The QuantiFluor® RNA system is applicable in many biological applications, like determining yield of in vitro transcribed RNA and measuring RNA concentration before Northern blot analysis, S1 nuclease assays, RNase protection assays, cDNA library preparation, RT-PCR and differential display PCR.
Fornecimento: Supplied with 20X TE buffer (pH 7,5), RNA standard and QuantiFluor® RNA dye.