Print…
Forceps, Chemware®
Fornecedor: Saint-Gobain
Chemware®
VERND1069288EA 24.4 EUR
VERND1069288
Forceps, Chemware®
Forceps
Forceps made from nickel-plated steel covered with an extra heavy-duty coating of PTFE.
  • For handling hot or chemically active materials
  • Non stick surface is inert
  • Coating prevents scratching glass and other delicate surfaces