- Inicial
- Padrões
- Biological Standards
- Peptide Reference Mix
Peptide Reference Mix
Fornecedor: Cayman Chemical
PROMV7491EA
317
EUR
PROMV7491
PROMV7495
Peptide Reference Mix
Padrões
Biological Standards
The 6×5 LC-MS/MS peptide reference mix is intended for use in method development, optimisation and for routine performance monitoring for optimal liquid chromatography (LC) performance, parameters like retention time, peak width and peak height and mass spectrometric (MS) parameters include mass accuracy, mass resolution, signal-tonoise, sensitivity, limit of detection (LOD), limit of quantitation (LOQ) and dynamic range.
- Peptide Mix and Free Software to Monitor LC/MS Instrument Performance
- Determine mass spec instrument sensitivity and dynamic range
- No manual calculations, maximised instrument performance with free PReMiS™ Software
The 6×5 LC-MS/MS Peptide Reference Mix is a unique reagent designed to monitor liquid chromatography (LC) and mass spectrometry (MS) instrument performance and assist in method development and optimization. The product is a mixture of 30 peptides; 6 sets of 5 isotopologues of the same peptide sequence. The isotopologues differ only by the number of stable, heavy-labeled amino acids incorporated into the sequence. The labels consist of uniform 13C and 15N atoms. Chromatographically, each of the isotopologues is indistinguishable; however, since they differ in mass, they are clearly resolved by mass spectrometry. The isotopologues of each peptide are present in a series of tenfold dilutions. This format allows assessment of instrument dynamic range and sensitivity from a single run. Peptides with a wide range of hydrophobicities were chosen to enable reporting of LC column performance. In addition, the peptides were chosen for maximal stability. Amino acids prone to artificial post-translational modification (i.e., methionine, asparagine, etc.) were excluded from the sequences. None of the peptides have internal lysines or arginines and will therefore not be affected by trypsin or Lys-C. In addition there is a mass separation of at least 4 Daltons between the isotopologues, so that even low-resolution instruments can distinguish the masses. PReMiS Software Tool: The 6×5 LC-MS/MS Peptide Reference Mix is accompanied by a complementary PReMiS Software tool (available by download) that reports on key liquid chromatography and mass spec parameters (see table). The parameter reports can be exported to CSV or saved as .pdf files. In addition to the general reporting feature, performance parameters can be tracked over time, allowing a clear assessment of trends to pinpoint poor performance and maintenance needs. For those laboratories that have multiple instruments, the ability to compare parameters across instruments will also be available. Thermo (.raw) and ABSCIEX (.wiff) formats are available for direct importing. Other vendor formats can be imported after conversion to .mzml format. Data reports are rapidly generated (usually in less than 2 minutes), with clear presentation of the XIC of all 30 masses available for immediate viewing.
The Mix is accompanied by a complementary PReMiS™ Software tool (available by download) that reports on key liquid chromatography and mass spec parameters. The parameter reports can be exported to CSV or saved as PDF files. Performance parameters can be tracked over time for a clear assessment of trends to pinpoint poor performance and maintenance needs. A function for comparing parameters across instruments are also available. Thermo (.raw) and ABSCIEX (.wiff) formats are available for direct importing. Other vendor formats can be imported after conversion to .mzml format. Data reports are usually generated in less than two minutes with clear presentation of the XIC of all 30 masses available for immediate viewing.
The isotopologues differ only by the number of stable, heavy-labeled amino acids incorporated into the sequence. The labels consist of uniform 13C and 15N atoms. Chromatographically, all of the isotopologues are indistinguishable; however, since they differ in mass, they are resolved by mass spectrometry. The isotopologues of each peptide are present in tenfold dilutions, which allow assessment of instrument dynamic range and sensitivity from a single run. Peptides with a wide range of hydrophobicities enable reporting of LC column performance and maximal stability. Amino acids prone to artificial post-translational modification (i.e., methionine, asparagine, and more) were excluded from the sequences. None of the peptides have internal lysines or arginines and are not affected by trypsin or Lys-C. There is also a mass separation of at least four Daltons between the isotopologues, so even low-resolution instruments can distinguish the masses.
Fornecimento: 6×5 LC-MS/MS peptide reference mix; aliquot tubes (6).
Novo produto
Promoção por tempo limitado
Os produtos selecionados estão em promoção
Inserir detalhes do certificado
Preço sob consulta
Learn more
About VWR
Item (s) adicionado (s) ao cesto